3-Hydroxy-2-methoxybenzaldehyde 98%
Dodavatel: Apollo Scientific
OR957483-1G
OR957483-250MG
OR957483-5G
APOSOR957483-1GEA
5920
CZK
APOSOR957483-1G
APOSOR957483-250MG
APOSOR957483-5G
3-Hydroxy-2-methoxybenzaldehyde 98%
3-Hydroxy-2-methoxybenzaldehyde
Vzorec:
C₈H₈O₃ MW: 152,147 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06656119 CAS číslo: 66495-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...