tert-Butyl 3-hydroxypropionate 95%
Dodavatel: Apollo Scientific
OR957444-1G
OR957444-5G
APOSOR957444-1GEA
4670
CZK
APOSOR957444-1G
APOSOR957444-5G
tert-Butyl 3-hydroxypropionate 95%
tert-Butyl 3-hydroxypropionate
Vzorec:
C₇H₁₄O₃ Storage Temperature: Ambient |
MDL Number:
MFCD04974574 CAS číslo: 59854-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...