Tisk…

1-(3-Chlorophenyl)-2-nitroethene 95%

Dodavatel: Apollo Scientific

OR957310-10G OR957310-1G OR957310-5G
APOSOR957310-10GEA 11380 CZK
APOSOR957310-10G APOSOR957310-1G APOSOR957310-5G
1-(3-Chlorophenyl)-2-nitroethene 95%
1-(3-Chlorophenyl)-2-nitroethene
Vzorec: C₈H₆ClNO₂
MW: 183,592 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00175518
CAS číslo: 3156-35-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR