Tisk…

(2-Chlorobenzyl)propylamine 95%

Dodavatel: Apollo Scientific

OR956996-1G OR956996-5G
APOSOR956996-1GEA 6220 CZK
APOSOR956996-1G APOSOR956996-5G
(2-Chlorobenzyl)propylamine 95%
(2-Chlorobenzyl)propylamine
Vzorec: C₁₀H₁₄ClN
MW: 183,678 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07405978
CAS číslo: 807343-03-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR