2-(Pyrrolidin-1-ylmethyl)cyclohexanone 95%
Dodavatel: Apollo Scientific
OR956501-1G
OR956501-250MG
APOSOR956501-1GEA
23580
CZK
APOSOR956501-1G
APOSOR956501-250MG
2-(Pyrrolidin-1-ylmethyl)cyclohexanone 95%
2-(Pyrrolidin-1-ylmethyl)cyclohexanone
Vzorec:
C₁₁H₁₉NO MW: 181,275 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16825284 CAS číslo: 82087-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...