Tisk…

5-Formyl-1H-pyrrole-2-carboxylic acid 98%

Dodavatel: Apollo Scientific

OR956120-1G OR956120-250MG
APOSOR956120-1GEA 2280 CZK
APOSOR956120-1G APOSOR956120-250MG
5-Formyl-1H-pyrrole-2-carboxylic acid 98%
5-Formyl-1H-pyrrole-2-carboxylic acid
Vzorec: C₆H₅NO₃
MW: 139,109 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09754206
CAS číslo: 7126-51-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR