Tisk…

Methyl 4-formyl-3-methylbenzoate 97%

Dodavatel: Apollo Scientific

OR955987-1G OR955987-250MG OR955987-5G
APOSOR955987-1GEA 4800 CZK
APOSOR955987-1G APOSOR955987-250MG APOSOR955987-5G
Methyl 4-formyl-3-methylbenzoate 97%
Methyl 4-formyl-3-methylbenzoate
Vzorec: C₁₀H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD20923636
CAS číslo: 24078-24-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR