Tisk…

Methyl 4-chloro-3-(chlorosulphonyl)benzoate 96%

Dodavatel: Apollo Scientific
Nebezpečí

OR955980-1G OR955980-250MG OR955980-5G
APOSOR955980-1GEA 9100 CZK
APOSOR955980-1G APOSOR955980-250MG APOSOR955980-5G
Methyl 4-chloro-3-(chlorosulphonyl)benzoate 96%
Methyl 4-chloro-3-(chlorosulphonyl)benzoate
Vzorec: C₈H₆Cl₂O₄S
MW: 269.102 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09816501
CAS číslo: 1000933-19-6
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR