Tisk…

Methyl 3-isopropoxybenzoate 95%

Dodavatel: Apollo Scientific

OR955972-1G OR955972-5G
APOSOR955972-1GEA 7670 CZK
APOSOR955972-1G APOSOR955972-5G
Methyl 3-isopropoxybenzoate 95%
Methyl 3-isopropoxybenzoate
Vzorec: C₁₁H₁₄O₃
MW: 194,227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01922057
CAS číslo: 350989-42-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR