Tisk…
Methyl 3-chloro-5-hydroxybenzoate 98%
Dodavatel: Apollo Scientific
OR955921-1G OR955921-250MG
APOSOR955921-1GEA 10650 CZK
APOSOR955921-1G APOSOR955921-250MG
Methyl 3-chloro-5-hydroxybenzoate 98%
Methyl 3-chloro-5-hydroxybenzoate
Vzorec: C₈H₇ClO₃
MW: 186.592 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12407235
CAS číslo: 98406-04-3