Methyl 2-amino-2-ethylbutanoate 96%
Dodavatel: Apollo Scientific
OR955916-1G
OR955916-250MG
APOSOR955916-1GEA
17990
CZK
APOSOR955916-1G
APOSOR955916-250MG
Methyl 2-amino-2-ethylbutanoate 96%
Methyl 2-amino-2-ethylbutanoate
Vzorec:
C₇H₁₅NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD12151684 CAS číslo: 70974-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...