Tisk…

Methyl-3-(4-pyridyl)propanoate 97%

Dodavatel: Apollo Scientific

Synonyms: Methyl 3-(4-Pyridyl)propanoate

OR955804-1G
APOSOR955804-1GEA 10250 CZK
APOSOR955804-1G
Methyl-3-(4-pyridyl)propanoate 97%
Methyl-3-(4-pyridyl)propanoate
Vzorec: C₉H₁₁NO₂
MW: 165,189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367499
CAS číslo: 90610-07-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR