(S)-1-(3-Methoxyphenyl)ethylamine hydrochloride 95%
Dodavatel: Apollo Scientific
OR955802-1G
OR955802-250MG
OR955802-5G
APOSOR955802-1GEA
7050
CZK
APOSOR955802-1G
APOSOR955802-250MG
APOSOR955802-5G
(S)-1-(3-Methoxyphenyl)ethylamine hydrochloride 95%
(S)-1-(3-Methoxyphenyl)ethylamine hydrochloride
Vzorec:
C₉H₁₃NO·ClH Storage Temperature: Ambient |
MDL Number:
MFCD11865390 CAS číslo: 1304771-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...