Tisk…

Methyl 2-(6-chloropyridin-3-yl)acetate 95%

Dodavatel: Apollo Scientific

OR955786-5G
APOSOR955786-5GEA 1620 CZK
APOSOR955786-5G
Methyl 2-(6-chloropyridin-3-yl)acetate 95%
Methyl 2-(6-chloropyridin-3-yl)acetate
Vzorec: C₈H₈ClNO₂
MW: 185,608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15526721
CAS číslo: 717106-69-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR