Tisk…

Methyl quinoline-8-carboxylate 95%

Dodavatel: Apollo Scientific

OR955684-1G OR955684-5G
APOSOR955684-1GEA 7870 CZK
APOSOR955684-1G APOSOR955684-5G
Methyl quinoline-8-carboxylate 95%
Methyl quinoline-8-carboxylate
Vzorec: C₁₁H₉NO₂
MW: 187,195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203774
CAS číslo: 40245-26-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR