Tisk…

Methyl 2-(2-bromo-5-chlorophenyl)acetate 97%

Dodavatel: Apollo Scientific

OR955643-5G
APOSOR955643-5GEA 2150 CZK
APOSOR955643-5G
Methyl 2-(2-bromo-5-chlorophenyl)acetate 97%
Methyl 2-(2-bromo-5-chlorophenyl)acetate
Vzorec: C₉H₈BrClO₂
MW: 263,516 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18398721
CAS číslo: 455957-76-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR