Methyl 4-Boc-morpholine-2-carboxylate 97%
Dodavatel: Apollo Scientific
Synonyms:
4-tert-Butyl 2-methyl morpholine-2,4-dicarboxylate
OR955607-1G
APOSOR955607-1GEA
2030
CZK
APOSOR955607-1G
Methyl 4-Boc-morpholine-2-carboxylate 97%
Methyl 4-Boc-morpholine-2-carboxylate
Vzorec:
C₁₁H₁₉NO₅ MW: 245,272 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16877230 CAS číslo: 500789-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...