Tisk…

3,4-Dimethyl pyridine-3,4-dicarboxylate 95%

Dodavatel: Apollo Scientific

OR955467-1G OR955467-5G
APOSOR955467-1GEA 3480 CZK
APOSOR955467-1G APOSOR955467-5G
3,4-Dimethyl pyridine-3,4-dicarboxylate 95%
3,4-Dimethyl pyridine-3,4-dicarboxylate
Vzorec: C₉H₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD00160293
CAS číslo: 1796-83-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR