Tisk…

Methyl (E)-3-(m-tolyl)prop-2-enoate 98%

Dodavatel: Apollo Scientific

OR955444-1G OR955444-5G
APOSOR955444-1GEA 6220 CZK
APOSOR955444-1G APOSOR955444-5G
Methyl (E)-3-(m-tolyl)prop-2-enoate 98%
Methyl (E)-3-(m-tolyl)prop-2-enoate
Vzorec: C₁₁H₁₂O₂
MW: 176,212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19441320
CAS číslo: 95416-56-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR