Tisk…

1-(Pyridin-3-yl)butan-1-one 95%

Dodavatel: Apollo Scientific

OR955084-1G OR955084-250MG
APOSOR955084-1GEA 3810 CZK
APOSOR955084-1G APOSOR955084-250MG
1-(Pyridin-3-yl)butan-1-one 95%
1-(Pyridin-3-yl)butan-1-one
Vzorec: C₉H₁₁NO
MW: 149,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12153276
CAS číslo: 1701-70-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR