Tisk…

2-Formyl-5-phenylphenol 96%

Dodavatel: Apollo Scientific

OR954913-1G OR954913-250MG
APOSOR954913-1GEA 10350 CZK
APOSOR954913-1G APOSOR954913-250MG
2-Formyl-5-phenylphenol 96%
2-Formyl-5-phenylphenol
Vzorec: C₁₃H₁₀O₂
MW: 198,217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18314600
CAS číslo: 35664-67-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR