Tisk…

3-Cyano-5-phenylphenol 98%

Dodavatel: Apollo Scientific

OR954905-1G OR954905-250MG
APOSOR954905-1GEA 10350 CZK
APOSOR954905-1G APOSOR954905-250MG
3-Cyano-5-phenylphenol 98%
3-Cyano-5-phenylphenol
Vzorec: C₁₃H₉NO
MW: 195,217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18314129
CAS číslo: 939771-50-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR