4-(Furan-2-yl)-2-methylphenol 96%
Dodavatel: Apollo Scientific
OR954884-1G
OR954884-250MG
APOSOR954884-1GEA
16340
CZK
APOSOR954884-1G
APOSOR954884-250MG
4-(Furan-2-yl)-2-methylphenol 96%
4-(Furan-2-yl)-2-methylphenol
Vzorec:
C₁₁H₁₀O₂ MW: 174,196 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17677675 CAS číslo: 1255638-84-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...