4-(2-Formylphenyl)phenol 95%
Dodavatel: Apollo Scientific
Synonyms:
2-(4-Hydroxyphenyl)benzaldehyde
OR954780-1G
OR954780-250MG
APOSOR954780-1GEA
15310
CZK
APOSOR954780-1G
APOSOR954780-250MG
4-(2-Formylphenyl)phenol 95%
4-(2-Formylphenyl)phenol
Vzorec:
C₁₃H₁₀O₂ MW: 198,217 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04117373 CAS číslo: 400747-55-9 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...