Tisk…

3-(4-Methylphenyl)phenol 98%

Dodavatel: Apollo Scientific

OR954770-1G OR954770-5G
APOSOR954770-1GEA 4770 CZK
APOSOR954770-1G APOSOR954770-5G
3-(4-Methylphenyl)phenol 98%
3-(4-Methylphenyl)phenol
Vzorec: C₁₃H₁₂O
MW: 184,234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06802317
CAS číslo: 486455-30-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR