Tisk…

6-(4-Cyanophenyl)-2-hydroxypyridine 98%

Dodavatel: Apollo Scientific

OR954658-1G OR954658-5G
APOSOR954658-1GEA 6220 CZK
APOSOR954658-1G APOSOR954658-5G
6-(4-Cyanophenyl)-2-hydroxypyridine 98%
6-(4-Cyanophenyl)-2-hydroxypyridine
Vzorec: C₁₂H₈N₂O
MW: 196,205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11876110
CAS číslo: 1111110-50-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR