7-Chloro-2-methyl-3H-quinazolin-4-one 98%
Dodavatel: Apollo Scientific
OR954613-1G
OR954613-25G
OR954613-5G
APOSOR954613-1GEA
4770
CZK
APOSOR954613-1G
APOSOR954613-25G
APOSOR954613-5G
7-Chloro-2-methyl-3H-quinazolin-4-one 98%
7-Chloro-2-methyl-3H-quinazolin-4-one
Vzorec:
C₉H₇ClN₂O MW: 194,618 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00844409 CAS číslo: 7012-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...