Tisk…

2-(4-Methoxyphenyl)-1,3,4-oxadiazole 95%

Dodavatel: Apollo Scientific

OR954531-1G OR954531-250MG
APOSOR954531-1GEA 8010 CZK
APOSOR954531-1G APOSOR954531-250MG
2-(4-Methoxyphenyl)-1,3,4-oxadiazole 95%
2-(4-Methoxyphenyl)-1,3,4-oxadiazole
Vzorec: C₉H₈N₂O₂
MW: 176,172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00491615
CAS číslo: 829-35-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR