4,4,8-Trimethyl-1,3-dihydroquinolin-2-one 98%
Dodavatel: Apollo Scientific
OR954468-10G
OR954468-1G
OR954468-5G
APOSOR954468-10GEA
14290
CZK
APOSOR954468-10G
APOSOR954468-1G
APOSOR954468-5G
4,4,8-Trimethyl-1,3-dihydroquinolin-2-one 98%
4,4,8-Trimethyl-1,3-dihydroquinolin-2-one
Vzorec:
C₁₂H₁₅NO MW: 189,254 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12913838 CAS číslo: 120689-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...