1-Butyl-1,3-benzodiazole-5-carbonitrile 98%
Dodavatel: Apollo Scientific
OR954261-1G
OR954261-25G
OR954261-5G
APOSOR954261-1GEA
4140
CZK
APOSOR954261-1G
APOSOR954261-25G
APOSOR954261-5G
1-Butyl-1,3-benzodiazole-5-carbonitrile 98%
1-Butyl-1,3-benzodiazole-5-carbonitrile
Vzorec:
C₁₂H₁₃N₃ MW: 199,252 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22549339 CAS číslo: 1403483-90-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...