Tisk…

6-Chloro-1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine 95%

Dodavatel: Apollo Scientific

OR954203-1G OR954203-250MG
APOSOR954203-1GEA 24170 CZK
APOSOR954203-1G APOSOR954203-250MG
6-Chloro-1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine 95%
6-Chloro-1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
Vzorec: C₆H₆ClN₅
MW: 183,598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18793420
CAS číslo: 5413-96-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR