Tisk…
Methyl 2-methylpiperidine-2-carboxylate 95%
Dodavatel: Apollo Scientific
OR954068-1G OR954068-250MG
APOSOR954068-1GEA 16340 CZK
APOSOR954068-1G APOSOR954068-250MG
Methyl 2-methylpiperidine-2-carboxylate 95%
Methyl 2-methylpiperidine-2-carboxylate
Vzorec: C₈H₁₅NO₂
MW: 157,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13179033
CAS číslo: 89115-93-5