4-Ethoxy-3-methoxybenzoic acid 98%
Dodavatel: Apollo Scientific
OR953981-10G
OR953981-1G
OR953981-25G
APOSOR953981-10GEA
11810
CZK
APOSOR953981-10G
APOSOR953981-1G
APOSOR953981-25G
4-Ethoxy-3-methoxybenzoic acid 98%
4-Ethoxy-3-methoxybenzoic acid
Vzorec:
C₁₀H₁₂O₄ MW: 196,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00127725 CAS číslo: 3535-30-6 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...