Tisk…

3-(2-Chlorophenoxy)propylamine 95%

Dodavatel: Apollo Scientific

OR953830-1G OR953830-250MG
APOSOR953830-1GEA 14290 CZK
APOSOR953830-1G APOSOR953830-250MG
3-(2-Chlorophenoxy)propylamine 95%
3-(2-Chlorophenoxy)propylamine
Vzorec: C₉H₁₂ClNO
MW: 185,651 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09883052
CAS číslo: 50911-59-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR