Tisk…

3-(2-Thienyl)-1-propanol 95%

Dodavatel: Apollo Scientific

OR953826-1G OR953826-5G
APOSOR953826-1GEA 2360 CZK
APOSOR953826-1G APOSOR953826-5G
3-(2-Thienyl)-1-propanol 95%
3-(2-Thienyl)-1-propanol
Vzorec: C₇H₁₀OS
Storage Temperature: Ambient
MDL Number: MFCD09927267
CAS číslo: 19498-72-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR