Tisk…

(E)-3-(Quinolin-2-yl)acrylic acid 96%

Dodavatel: Apollo Scientific

OR953796-1G OR953796-250MG OR953796-5G
APOSOR953796-1GEA 8700 CZK
APOSOR953796-1G APOSOR953796-250MG APOSOR953796-5G
(E)-3-(Quinolin-2-yl)acrylic acid 96%
(E)-3-(Quinolin-2-yl)acrylic acid
Vzorec: C₁₂H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00182035
CAS číslo: 81124-50-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR