Tisk…

2-Thienyl-carbamic acid ethyl ester 95%

Dodavatel: Apollo Scientific

OR953708-1G OR953708-250MG
APOSOR953708-1GEA 22950 CZK
APOSOR953708-1G APOSOR953708-250MG
2-Thienyl-carbamic acid ethyl ester 95%
2-Thienyl-carbamic acid ethyl ester
Vzorec: C₇H₉NO₂S
MW: 171,217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568164
CAS číslo: 105995-16-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR