2-(5-Oxazolyl)benzoic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR953704-1G
OR953704-250MG
APOSOR953704-1GEA
8070
CZK
APOSOR953704-1G
APOSOR953704-250MG
2-(5-Oxazolyl)benzoic acid 95%
2-(5-Oxazolyl)benzoic acid
Vzorec:
C₁₀H₇NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD18375277 CAS číslo: 169508-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...