Methyl 2-furoylacetate 95%
Dodavatel: Apollo Scientific
OR953640-1G
OR953640-5G
APOSOR953640-1GEA
2690
CZK
APOSOR953640-1G
APOSOR953640-5G
Methyl 2-furoylacetate 95%
Methyl 2-furoylacetate
Vzorec:
C₈H₈O₄ Storage Temperature: Ambient |
MDL Number:
MFCD03424758 CAS číslo: 615-06-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...