Tisk…

2-Chloroallyl alcohol

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Chloro-3-hydroxyprop-1-ene, 2-Chloroprop-2-en-1-ol

OR9536-100G OR9536-25G OR9536-5G
APOSOR9536-100GEA 18850 CZK
APOSOR9536-100G APOSOR9536-25G APOSOR9536-5G
2-Chloroallyl alcohol
2-Chloroallyl alcohol
Vzorec: C₃H₅ClO
MW: 92,524 g/mol
Bod varu: 133…134 °C
Hustota: 1,162 g/cm³ (25 °C)
Bod vzplanutí: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00004701
CAS číslo: 5976-47-6
UN: 2920
ADR: 8+3,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR