(2-Hydroxyphenyl)acetonitrile 95%
Dodavatel: Apollo Scientific
OR953586-1G
OR953586-250MG
OR953586-5G
APOSOR953586-1GEA
3310
CZK
APOSOR953586-1G
APOSOR953586-250MG
APOSOR953586-5G
(2-Hydroxyphenyl)acetonitrile 95%
(2-Hydroxyphenyl)acetonitrile
Vzorec:
C₈H₇NO Storage Temperature: Ambient |
MDL Number:
MFCD00128130 CAS číslo: 14714-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...