Tisk…

Ethyl 2-(4-hydroxypyrimidin-5-yl)acetate 95%

Dodavatel: Apollo Scientific

OR953529-1G OR953529-250MG
APOSOR953529-1GEA 4070 CZK
APOSOR953529-1G APOSOR953529-250MG
Ethyl 2-(4-hydroxypyrimidin-5-yl)acetate 95%
Ethyl 2-(4-hydroxypyrimidin-5-yl)acetate
Vzorec: C₈H₁₀N₂O₃
MW: 182,177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11656636
CAS číslo: 6214-46-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR