Tisk…

2-(Cyclohexyloxy)ethylamine hydrochloride 95%

Dodavatel: Apollo Scientific

OR953493-1G OR953493-250MG
APOSOR953493-1GEA 9330 CZK
APOSOR953493-1G APOSOR953493-250MG
2-(Cyclohexyloxy)ethylamine hydrochloride 95%
2-(Cyclohexyloxy)ethylamine hydrochloride
Vzorec: C₈H₁₇NO·ClH
MW: 179,688 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12913027
CAS číslo: 1193387-70-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR