Tisk…

4-(1H-Imidazol-2-yl)benzoic acid 96%

Dodavatel: Apollo Scientific

OR953354-5G
APOSOR953354-5GEA 2730 CZK
APOSOR953354-5G
4-(1H-Imidazol-2-yl)benzoic acid 96%
4-(1H-Imidazol-2-yl)benzoic acid
Vzorec: C₁₀H₈N₂O₂
MW: 188,183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00456057
CAS číslo: 108035-45-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR