2-Amino-3-phenylpropanenitrile hydrochloride 95%
Dodavatel: Apollo Scientific
OR953337-1G
OR953337-250MG
APOSOR953337-1GEA
17160
CZK
APOSOR953337-1G
APOSOR953337-250MG
2-Amino-3-phenylpropanenitrile hydrochloride 95%
2-Amino-3-phenylpropanenitrile hydrochloride
Vzorec:
C₉H₁₀N₂·ClH MW: 182.65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00082676 CAS číslo: 93554-83-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...