Tisk…

(S)-b-Amino-cyclohexanepropanol 97%

Dodavatel: Apollo Scientific

OR953185-1G OR953185-250MG OR953185-500MG
APOSOR953185-1GEA 20470 CZK
APOSOR953185-1G APOSOR953185-250MG APOSOR953185-500MG
(S)-b-Amino-cyclohexanepropanol 97%
(S)-b-Amino-cyclohexanepropanol
Vzorec: C₉H₁₉NO
Storage Temperature: Ambient
MDL Number: MFCD00191731
CAS číslo: 131288-67-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR