3-Chloro-1-benzothiophene-2-carbaldehyde 98%
Dodavatel: Apollo Scientific
OR953129-10G
OR953129-1G
OR953129-5G
APOSOR953129-10GEA
28540
CZK
APOSOR953129-10G
APOSOR953129-1G
APOSOR953129-5G
3-Chloro-1-benzothiophene-2-carbaldehyde 98%
3-Chloro-1-benzothiophene-2-carbaldehyde
Vzorec:
C₉H₅ClOS MW: 196,653 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00489051 CAS číslo: 14006-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...