Tisk…
Methyl 2-(2-pyrimidyl)acetate 98%
Dodavatel: Apollo Scientific
Synonyms: Methyl 2-(2-Pyrimidyl)acetate
OR953099-1G
APOSOR953099-1GEA 1870 CZK
APOSOR953099-1G
Methyl 2-(2-pyrimidyl)acetate 98%
Methyl 2-(2-pyrimidyl)acetate
Vzorec: C₇H₈N₂O₂
MW: 152,151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12198887
CAS číslo: 60561-50-4