Tisk…

2-(Cyclopentyloxy)ethylamine 95%

Dodavatel: Apollo Scientific

OR953041-1G
APOSOR953041-1GEA 5790 CZK
APOSOR953041-1G
2-(Cyclopentyloxy)ethylamine 95%
2-(Cyclopentyloxy)ethylamine
Vzorec: C₇H₁₅NO
MW: 129,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09730285
CAS číslo: 933732-12-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR