Tisk…
cis-4-Methyl-cyclohexylamine hydrochloride 95%
Dodavatel: Apollo Scientific
Synonyms: cis-4-Methylcyclohexanamine hydrochloride
OR953018-100MG OR953018-1G OR953018-250MG
APOSOR953018-100MGEA 1410 CZK
APOSOR953018-100MG APOSOR953018-1G APOSOR953018-250MG
cis-4-Methyl-cyclohexylamine hydrochloride 95%
cis-4-Methyl-cyclohexylamine hydrochloride
Vzorec: C₇H₁₅N·ClH
MW: 149,662 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09991703
CAS číslo: 33483-66-8